Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKS1B SIRNA Assays ›
Gene Symbol:
ANKS1B
Gene Name:
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases:
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location:
Chr.12: 98729904 - 99984727 on Build GRCh38
UniGene ID:
Hs.506458
Species:
Human
siRNA ID
HSS183592
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKS1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204066.1
NP_001190995.1
3
202
NM_001204067.1
NP_001190996.1
3
263
NM_001204068.1
NP_001190997.1
2
546
NM_001204069.1
NP_001190998.1
2
546
NM_001204070.1
NP_001190999.1
2
546
NM_020140.3
NP_064525.1
2
546
NM_152788.4
NP_690001.3
16
2978
NM_181670.3
NP_858056.2
2
546
XM_005269028.4
XP_005269085.1
16
2589
XM_005269029.4
XP_005269086.1
16
2549
XM_006719504.3
XP_006719567.1
16
3051
XM_006719505.3
XP_006719568.1
16
3051
XM_006719506.3
XP_006719569.1
16
3051
XM_006719507.3
XP_006719570.1
16
3051
XM_006719508.3
XP_006719571.1
16
3051
XM_006719509.3
XP_006719572.1
16
3051
XM_006719510.3
XP_006719573.1
16
3051
XM_006719512.3
XP_006719575.1
16
3051
XM_006719513.3
XP_006719576.1
16
3051
XM_006719514.3
XP_006719577.1
16
3039
XM_011538571.2
XP_011536873.1
16
3051
XM_017019651.1
XP_016875140.1
16
3051
XM_017019652.1
XP_016875141.1
16
3039
XM_017019653.1
XP_016875142.1
16
3051
XM_017019654.1
XP_016875143.1
16
3039
XM_017019655.1
XP_016875144.1
16
3051
XM_017019656.1
XP_016875145.1
16
3051
XM_017019657.1
XP_016875146.1
16
3051
XM_017019658.1
XP_016875147.1
16
3051
XM_017019659.1
XP_016875148.1
2
542
XM_017019660.1
XP_016875149.1
2
542
XM_017019661.1
XP_016875150.1
3
256
XM_017019662.1
XP_016875151.1
3
386
XM_017019663.1
XP_016875152.1
2
542
XM_017019664.1
XP_016875153.1
3
198
XM_017019665.1
XP_016875154.1
2
542
GenBank mRNA
AF145204.1
-
10
1682
AF164792.1
-
2
479
AK289768.1
-
2
511
AK294191.1
-
3
263
AY281131.1
-
16
2564
AY281132.1
-
2
239
AY283057.2
-
9
1595
AY620824.1
-
7
1289
AY753193.1
-
2
239
BC026313.1
-
2
441
BC068451.1
-
3
202
BC150204.1
-
2
493
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of synaptic plasticity by receptor localization to synapse
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-nglqg:80/10.239.166.193:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0