Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130725.1 | NP_001124197.1 | 3 | 667 | ||
NM_001144932.2 | NP_001138404.1 | 4 | 1105 | |||
NM_002797.4 | NP_002788.1 | 3 | 1020 | |||
GenBank mRNA | AK225888.1 | - | 3 | 785 | ||
AK300714.1 | - | 4 | 850 | |||
AK311895.1 | - | 3 | 776 | |||
BC057840.1 | - | 3 | 775 | 4795732 | ||
BC107720.1 | - | 3 | 771 | 6738409 | ||
BT006777.1 | - | 3 | 591 | |||
BU539357.1 | - | 3 | 643 | |||
BU632351.1 | - | 1 | 288 | |||
BX248299.1 | - | 4 | 837 | |||
BX538001.1 | - | 3 | 767 | |||
CD048996.1 | - | 3 | 699 | |||
S74378.1 | - | 3 | 612 |