Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300743.1 | NP_001287672.1 | 17 | 1720 | ||
NM_020314.5 | NP_064710.4 | 19 | 1921 | |||
XM_005255435.3 | XP_005255492.1 | 19 | 1873 | |||
XM_006721065.3 | XP_006721128.1 | 17 | 1672 | |||
XM_011545896.2 | XP_011544198.1 | 19 | 1622 | |||
XM_011545897.2 | XP_011544199.1 | 12 | 989 | |||
GenBank mRNA | AF461052.2 | - | 13 | 1417 | ||
AK024693.1 | - | 15 | 1191 | |||
AK095078.1 | - | 17 | 1417 | |||
AK290286.1 | - | 19 | 1618 | |||
AL136744.1 | - | 6 | 534 | |||
AL832159.1 | - | 19 | 1617 | |||
BC014900.2 | - | 18 | 1589 | 3909068 | ||
BC050464.1 | - | 19 | 1661 | 6061139 | ||
BC058845.1 | - | 19 | 1620 | 6452778 | ||
BX537867.1 | - | 19 | 1633 |