Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040194.1 | NP_001035284.1 | 4 | 336 | ||
NM_001040195.1 | NP_001035285.1 | 3 | 301 | |||
NM_001040196.1 | NP_001035286.1 | 5 | 433 | |||
NM_001040197.1 | NP_001035287.1 | 5 | 433 | |||
NM_020350.4 | NP_065083.3 | 4 | 336 | |||
XM_011541799.2 | XP_011540101.2 | 5 | 350 | |||
XM_011541800.2 | XP_011540102.1 | 5 | 350 | |||
XM_011541802.1 | XP_011540104.1 | 6 | 447 | |||
GenBank mRNA | AF165187.1 | - | 4 | 212 | ||
AF312374.1 | - | 4 | 267 | |||
AK290042.1 | - | 4 | 279 | |||
BC017328.2 | - | 4 | 274 | 4546585 | ||
BE782705.1 | - | 386 | ||||
BI117521.1 | - | 4 | 308 | |||
CA866314.1 | - | 3 | 307 | |||
CD674964.1 | - | 5 | 313 |