Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PCNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320395.1 | NP_001307324.1 | 2 | 299 | ||
NM_001320397.1 | NP_001307326.1 | 2 | 299 | |||
NM_001320398.1 | NP_001307327.1 | 2 | 299 | |||
NM_001320399.1 | NP_001307328.1 | 3 | 425 | |||
NM_001320400.1 | NP_001307329.1 | 2 | 299 | |||
NM_001320401.1 | NP_001307330.1 | 2 | 299 | |||
NM_020357.2 | NP_065090.1 | 2 | 299 | |||
NR_135223.1 | 2 | 247 | ||||
NR_135225.1 | 2 | 247 | ||||
NR_135226.1 | 2 | 247 | ||||
NR_135227.1 | 2 | 500 | ||||
NR_135228.1 | 2 | 247 | ||||
NR_135230.1 | 2 | 247 | ||||
NR_135232.1 | 3 | 342 | ||||
XM_017006922.1 | XP_016862411.1 | 2 | 213 | |||
GenBank mRNA | AB037675.1 | - | 2 | 195 | ||
AK025551.1 | - | 2 | 139 | |||
AK098760.1 | - | 2 | 250 | |||
AK222978.1 | - | 2 | 190 | |||
AK314629.1 | - | 2 | 180 | |||
BC013916.1 | - | 2 | 207 | 3929908 | ||
BC022001.2 | - | 2 | 197 | 4823990 | ||
BC107796.1 | - | 2 | 141 | |||
BF244508.1 | - | 191 | ||||
BI547939.1 | - | 2 | 412 | |||
CD696085.1 | - | 2 | 214 | |||
CN482876.1 | - | 3 | 226 | |||
DA041324.1 | - | 2 | 155 | |||
DA304502.1 | - | 2 | 247 | |||
DA710370.1 | - | 2 | 189 | |||
DA817951.1 | - | 2 | 167 | |||
DB060329.1 | - | 2 | 210 | |||
DB481581.1 | - | 2 | 135 |