Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079684.2 | NP_001073152.1 | 6 | 827 | ||
NM_001079685.2 | NP_001073153.1 | 6 | 806 | |||
NM_001287335.1 | NP_001274264.1 | 4 | 540 | |||
NM_001287336.1 | NP_001274265.1 | 5 | 719 | |||
NM_020444.4 | NP_065177.2 | 7 | 914 | |||
NR_109796.1 | 3 | 408 | ||||
NR_109797.1 | 3 | 375 | ||||
NR_109798.1 | 4 | 521 | ||||
NR_109799.1 | 4 | 483 | ||||
NR_109800.1 | 3 | 396 | ||||
XM_005265941.1 | XP_005265998.1 | 6 | 727 | |||
XM_005265945.1 | XP_005266002.1 | 4 | 402 | |||
XM_005265946.4 | XP_005266003.1 | 6 | 822 | |||
XM_011534595.1 | XP_011532897.1 | 6 | 689 | |||
XM_011534596.1 | XP_011532898.1 | 5 | 638 | |||
XM_017009651.1 | XP_016865140.1 | 5 | 623 | |||
XM_017009652.1 | XP_016865141.1 | 2 | 346 | |||
XM_017009653.1 | XP_016865142.1 | 4 | 495 | |||
GenBank mRNA | AK315428.1 | - | 7 | 871 | ||
AL390217.1 | - | 6 | 785 | |||
AL832558.1 | - | 1 | 637 | |||
BC006316.2 | - | 3 | 328 | |||
BC010448.1 | - | 7 | 849 | 4155679 | ||
BC012848.1 | - | 4 | 418 | |||
BC017097.1 | - | 7 | 871 | 3529175 | ||
BM547296.1 | - | 3 | 356 | |||
BX648859.1 | - | 7 | 913 | |||
CN341192.1 | - | 4 | 490 | |||
CR457331.1 | - | 5 | 541 | |||
DQ073392.1 | - | 5 | 541 | |||
HY120613.1 | - | 3 | 395 |