Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288951.1 | NP_001275880.1 | 15, 16 | 2161 | ||
NM_001288953.1 | NP_001275882.1 | 16, 17 | 2152 | |||
NM_001288955.1 | NP_001275884.1 | 14, 15 | 2003 | |||
NM_020458.3 | NP_065191.2 | 15, 16 | 2161 | |||
XM_005264439.3 | XP_005264496.1 | 16, 17 | 2314 | |||
XM_011532998.2 | XP_011531300.1 | 17, 18 | 2379 | |||
XM_011532999.2 | XP_011531301.1 | 15, 16 | 2104 | |||
XM_011533000.2 | XP_011531302.1 | 12, 13 | 1236 | |||
XM_011533001.2 | XP_011531303.1 | 8, 9 | 1026 | |||
XM_017004525.1 | XP_016860014.1 | 15, 16 | 1646 | |||
XM_017004526.1 | XP_016860015.1 | 12, 13 | 1856 | |||
XM_017004527.1 | XP_016860016.1 | 15, 16 | 2256 | |||
XM_017004528.1 | XP_016860017.1 | 16, 17 | 2321 | |||
XM_017004529.1 | XP_016860018.1 | 15, 16 | 2104 | |||
GenBank mRNA | AB032966.2 | - | 16, 17 | 1741 | ||
AK056464.1 | - | 16, 17 | 2124 | |||
AL834383.1 | - | 15, 16 | 2161 | |||
BC001978.1 | - | 6, 7 | 570 | |||
BC065554.1 | - | 12, 13 | 1279 | |||
BC082965.1 | - | 7, 8 | 900 | |||
BC111487.1 | - | 15, 16 | 2049 | 6041916 |