Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077203.2 | NP_001070671.1 | 9 | 1345 | ||
NM_001282801.1 | NP_001269730.1 | 9 | 1342 | |||
NM_001282802.1 | NP_001269731.1 | 9 | 1490 | |||
NM_001282803.1 | NP_001269732.1 | 7 | 1097 | |||
NM_020654.4 | NP_065705.3 | 10 | 1540 | |||
XM_005247662.4 | XP_005247719.1 | 8 | 1133 | |||
XM_011513038.2 | XP_011511340.1 | 10 | 1529 | |||
XM_011513039.2 | XP_011511341.1 | 9 | 1454 | |||
XM_011513040.2 | XP_011511342.1 | 9 | 1334 | |||
XM_011513041.2 | XP_011511343.1 | 9 | 1331 | |||
XM_011513042.2 | XP_011511344.1 | 8 | 1136 | |||
XM_011513045.2 | XP_011511347.1 | 10 | 1530 | |||
XM_017006924.1 | XP_016862413.1 | 10 | 1417 | |||
XM_017006925.1 | XP_016862414.1 | 10 | 1505 | |||
XM_017006926.1 | XP_016862415.1 | 8 | 1243 | |||
XM_017006927.1 | XP_016862416.1 | 8 | 1240 | |||
XM_017006928.1 | XP_016862417.1 | 10 | 1530 | |||
GenBank mRNA | AB051494.1 | - | 3 | 547 | ||
AF199458.1 | - | 9 | 1488 | |||
AK297356.1 | - | 1451 | ||||
AL136599.1 | - | 9 | 1368 | |||
AL833577.1 | - | 9 | 1311 | |||
BC129988.1 | - | 10 | 1467 | 40128878 | ||
BX537943.1 | - | 7 | 1041 |