Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCAML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020693.3 | NP_065744.2 | 12 | 2585 | ||
XM_011542917.1 | XP_011541219.1 | 12 | 2615 | |||
XM_011542918.1 | XP_011541220.1 | 13 | 2303 | |||
XM_011542919.2 | XP_011541221.1 | 7 | 1258 | |||
XM_011542920.2 | XP_011541222.1 | 7 | 1305 | |||
XM_011542921.2 | XP_011541223.1 | 3 | 862 | |||
XM_011542924.2 | XP_011541226.1 | 2 | 780 | |||
XM_011542925.2 | XP_011541227.1 | 2 | 1454 | |||
GenBank mRNA | AB032958.2 | - | 12 | 2520 | ||
AF304304.1 | - | 12 | 2415 | |||
AF304305.1 | - | 10 | 1782 | |||
AF334384.1 | - | 12 | 2497 | |||
AF491813.1 | - | 12 | 2585 |
Set Membership: |
![]() |