Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMKLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297776.1 | NP_001284705.1 | 6 | 1316 | ||
NM_020734.3 | NP_065785.2 | 6 | 1748 | |||
XM_006719118.2 | XP_006719181.1 | 6 | 1437 | |||
XM_011520776.1 | XP_011519078.1 | 6 | 1437 | |||
XM_011520777.1 | XP_011519079.1 | 6 | 1437 | |||
XM_011520778.2 | XP_011519080.1 | 6 | 1437 | |||
XM_011520779.2 | XP_011519081.1 | 6 | 1437 | |||
XM_017019677.1 | XP_016875166.1 | 6 | 1437 | |||
XM_017019678.1 | XP_016875167.1 | 6 | 1437 | |||
XM_017019679.1 | XP_016875168.1 | 6 | 1437 | |||
XM_017019682.1 | XP_016875171.1 | 6 | 1316 | |||
XM_017019683.1 | XP_016875172.1 | 7 | 2124 | |||
XM_017019684.1 | XP_016875173.1 | 6 | 1060 | |||
XM_017019685.1 | XP_016875174.1 | 7 | 1839 | |||
XM_017019686.1 | XP_016875175.1 | 6 | 1074 | |||
XM_017019687.1 | XP_016875176.1 | 7 | 1438 | |||
XM_017019688.1 | XP_016875177.1 | 6 | 1578 | |||
XM_017019689.1 | XP_016875178.1 | 6 | 1127 | |||
XM_017019690.1 | XP_016875179.1 | 6 | 938 | |||
GenBank mRNA | AB033064.1 | - | 6 | 1687 | ||
AK302847.1 | - | 6 | 1316 | |||
BC021977.2 | - | 3 | 412 |