Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080522.2 | NP_001073991.2 | 16 | 1878 | ||
XM_005248177.1 | XP_005248234.1 | 15 | 1833 | |||
XM_011513872.2 | XP_011512174.1 | 15 | 1833 | |||
XM_017008482.1 | XP_016863971.1 | 13 | 1627 | |||
GenBank mRNA | AB037766.1 | - | 13 | 1616 | ||
AK023876.1 | - | 7 | 788 | |||
AK096047.1 | - | 15 | 1817 | |||
AK309178.1 | - | 15 | 1808 | |||
BX647334.1 | - | 15 | 1813 | |||
EU450799.1 | - | 16 | 1705 |