Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFYVE28 SIRNA Assays ›
Gene Symbol:
ZFYVE28
Gene Name:
zinc finger FYVE-type containing 28
Gene Aliases:
LST2, LYST2
Chromosome Location:
Chr.4: 2269585 - 2418698 on Build GRCh38
UniGene ID:
Hs.292056
Species:
Human
siRNA ID
HSS184047
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFYVE28
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001172656.1
NP_001166127.1
4
665
NM_001172657.1
NP_001166128.1
4
665
NM_001172658.1
NP_001166129.1
3
524
NM_001172659.1
NP_001166130.1
4
698
NM_001172660.1
NP_001166131.1
4
698
NM_020972.2
NP_066023.2
4
665
XM_006713900.3
XP_006713963.1
4
719
XM_006713902.3
XP_006713965.1
4
720
XM_006713904.3
XP_006713967.1
4
719
XM_011513540.2
XP_011511842.1
2
133
XM_011513541.2
XP_011511843.1
4
719
XM_017008502.1
XP_016863991.1
4
2158
XM_017008503.1
XP_016863992.1
4
720
XM_017008504.1
XP_016863993.1
4
719
XM_017008505.1
XP_016863994.1
3
2263
XM_017008506.1
XP_016863995.1
4
720
XM_017008507.1
XP_016863996.1
4
719
XM_017008508.1
XP_016863997.1
4
719
XM_017008509.1
XP_016863998.1
4
719
XM_017008510.1
XP_016863999.1
4
719
GenBank mRNA
AB046863.1
-
4
664
AK126692.1
-
2
1530
AK293775.1
-
2
148
AK294710.1
-
5
492
AK294874.1
-
4
626
AK316484.1
-
4
698
BC053664.1
-
4
631
BC137309.1
-
5
408
9021306
BC137310.1
-
5
408
9021307
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of epidermal growth factor-activated receptor activity
negative regulation of epidermal growth factor receptor signaling pathway
protein binding
phosphatidylinositol-3-phosphate binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0