Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102402.2 | NP_001095872.1 | 5, 6 | 814 | ||
NM_021213.3 | NP_067036.2 | 5, 6 | 707 | |||
GenBank mRNA | AF087857.1 | - | 5, 6 | 692 | ||
AF114430.1 | - | 5, 6 | 597 | |||
AF151638.1 | - | 5, 6 | 691 | |||
AK024667.1 | - | 5, 6 | 633 | |||
AK058120.1 | - | 5, 6 | 724 | |||
AK314509.1 | - | 5, 6 | 633 | |||
BC005112.1 | - | 5, 6 | 519 | |||
BC012084.1 | - | 5, 6 | 625 | 4575824 | ||
BX648199.1 | - | 3, 4 | 2582 |