Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRODH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021232.1 | NP_067055.1 | 4 | 743 | ||
XM_017027084.1 | XP_016882573.1 | 3 | 600 | |||
XM_017027085.1 | XP_016882574.1 | 3 | 599 | |||
GenBank mRNA | BC039710.1 | - | 3 | 515 | ||
U80018.1 | - | 4 | 743 |