Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278622.1 | NP_001265551.1 | 13 | 1771 | ||
NM_002873.1 | NP_002864.1 | 12 | 1876 | |||
NM_133338.2 | NP_579916.1 | 14 | 1772 | |||
NM_133339.2 | NP_579917.1 | 11 | 1329 | |||
NM_133340.2 | NP_579918.1 | 11 | 1362 | |||
NM_133341.2 | NP_579919.1 | 11 | 1288 | |||
NM_133342.2 | NP_579920.1 | 13 | 1749 | |||
NM_133343.1 | NP_579921.1 | 13 | 1714 | |||
NM_133344.2 | NP_579922.1 | 12 | 1531 | |||
XM_017009679.1 | XP_016865168.1 | 13 | 1591 | |||
XM_017009680.1 | XP_016865169.1 | 12 | 1645 | |||
XM_017009681.1 | XP_016865170.1 | 12 | 1503 | |||
GenBank mRNA | AF017748.1 | - | 12 | 1501 | ||
AF076838.1 | - | 12 | 1876 | |||
AF085736.2 | - | 11 | 1329 | |||
AF098533.1 | - | 11 | 1308 | |||
AF098534.1 | - | 11 | 1234 | |||
AF112263.1 | - | 11 | 1353 | |||
AF126424.1 | - | 14 | 1771 | |||
AJ001642.1 | - | 12 | 1540 | |||
AJ004977.1 | - | 12 | 1536 | |||
AK292487.1 | - | 13 | 1749 | |||
AL122068.1 | - | 13 | 1714 | |||
BC032304.1 | - | 11 | 1342 | 5170876 | ||
BX537441.1 | - | 13 | 1771 |