Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021626.2 | NP_067639.1 | 12 | 1249 | ||
XM_005257557.2 | XP_005257614.1 | 11 | 1117 | |||
GenBank mRNA | AF113214.1 | - | 12 | 1174 | ||
AF265441.1 | - | 8 | 718 | |||
AF282618.1 | - | 12 | 1228 | |||
AK027373.1 | - | 12 | 1222 | |||
AK298161.1 | - | 12 | 1168 | |||
AY358559.1 | - | 12 | 1229 | |||
BC072405.1 | - | 12 | 1199 | 6137668 |