Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002917.1 | NP_002908.1 | 4, 5 | 580 | ||
XM_011523587.2 | XP_011521889.1 | 3, 4 | 274 | |||
GenBank mRNA | AF038196.1 | - | 1, 2 | 154 | ||
AK293568.1 | - | 5, 6 | 526 | |||
AK302068.1 | - | 4, 5 | 389 | |||
U94353.1 | - | 1, 2 | 152 |