Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199551.1 | NP_001186480.1 | 6 | 500 | ||
NM_001199552.1 | NP_001186481.1 | 5 | 470 | |||
NM_001199623.1 | NP_001186552.1 | 5 | 473 | |||
NM_001321142.1 | NP_001308071.1 | 6 | 583 | |||
NM_001321143.1 | NP_001308072.1 | 5 | 429 | |||
NM_001321144.1 | NP_001308073.1 | 4 | 316 | |||
NM_022094.3 | NP_071377.2 | 5 | 554 | |||
GenBank mRNA | AF303893.1 | - | 5 | 554 | ||
AK024524.1 | - | 4 | 313 | |||
AK024530.1 | - | 4 | 313 | |||
AL598630.1 | - | 5 | 474 | |||
AY364640.1 | - | 4 | 400 | |||
BC016851.1 | - | 4 | 449 | 3851667 | ||
BQ695178.1 | - | 6 | 569 | |||
CD518729.1 | - | 5 | 479 | |||
CD518776.1 | - | 5 | 555 | |||
CD519587.1 | - | 4 | 449 |
Gene Symbol: CIDECP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002786.1 | 2 | 144 | |||
GenBank mRNA | AF230335.1 | - | 2 | 355 | ||
BC006266.2 | - | 2 | 334 | |||
BC043599.1 | - | 2 | 365 |