Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199551.1 | NP_001186480.1 | 7 | 694 | ||
NM_001199552.1 | NP_001186481.1 | 6 | 664 | |||
NM_001199623.1 | NP_001186552.1 | 6 | 667 | |||
NM_001321142.1 | NP_001308071.1 | 7 | 777 | |||
NM_001321143.1 | NP_001308072.1 | 6 | 623 | |||
NM_001321144.1 | NP_001308073.1 | 5 | 510 | |||
NM_022094.3 | NP_071377.2 | 6 | 748 | |||
GenBank mRNA | AF303893.1 | - | 6 | 748 | ||
AI460316.1 | - | 1 | 535 | |||
AK024524.1 | - | 5 | 507 | |||
AK024530.1 | - | 5 | 507 | |||
AY364638.1 | - | 3 | 263 | |||
AY364640.1 | - | 5 | 594 | |||
BC016851.1 | - | 5 | 643 | 3851667 | ||
CD518776.1 | - | 6 | 749 |
Gene Symbol: CIDECP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF230335.1 | - | 3 | 549 |