Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040424.2 | NP_001035514.1 | 5 | 557 | ||
NM_001282934.1 | NP_001269863.1 | 5 | 557 | |||
NM_022115.4 | NP_071398.3 | 12 | 1544 | |||
NR_104257.1 | 5 | 557 | ||||
NR_104258.1 | 5 | 557 | ||||
NR_104260.1 | 5 | 412 | ||||
NR_135464.1 | 5 | 553 | ||||
XM_006724039.2 | XP_006724102.1 | 5 | 471 | |||
XM_006724040.2 | XP_006724103.1 | 5 | 471 | |||
XM_011529674.1 | XP_011527976.1 | 5 | 471 | |||
XM_011529675.1 | XP_011527977.1 | 5 | 413 | |||
XM_011529676.2 | XP_011527978.1 | 5 | 2822 | |||
XM_011529677.2 | XP_011527979.1 | 5 | 498 | |||
XM_011529678.2 | XP_011527980.1 | 5 | 542 | |||
XM_011529679.2 | XP_011527981.1 | 5 | 875 | |||
XM_011529681.2 | XP_011527983.1 | 5 | 1779 | |||
XM_011529683.1 | XP_011527985.1 | 11 | 1260 | |||
XM_017028425.1 | XP_016883914.1 | 5 | 412 | |||
XM_017028426.1 | XP_016883915.1 | 5 | 2821 | |||
GenBank mRNA | AB051812.1 | - | 5 | 548 | ||
AB051813.1 | - | 5 | 548 | |||
AB051814.1 | - | 5 | 548 | |||
AB051815.1 | - | 5 | 548 | |||
AF276513.1 | - | 12 | 1544 | |||
AY078498.2 | - | 12 | 1544 | |||
BC067102.1 | - | 5 | 412 | 5925140 | ||
BM544857.1 | - | 374 | ||||
HY003319.1 | - | 5 | 410 |