Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM110C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077710.2 | NP_001071178.2 | 1 | 1075 | ||
XM_011510372.2 | XP_011508674.1 | 1 | 1077 | |||
XM_017004689.1 | XP_016860178.1 | 2 | 1269 | |||
XM_017004690.1 | XP_016860179.1 | 2 | 1017 | |||
XM_017004691.1 | XP_016860180.1 | 1 | 1077 | |||
XM_017004692.1 | XP_016860181.1 | 1 | 1077 | |||
GenBank mRNA | AK302544.1 | - | 1 | 1016 | ||
DQ431183.1 | - | 1 | 872 |