Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM196A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039762.2 | NP_001034851.1 | 4 | 1291 | ||
XM_005252694.3 | XP_005252751.1 | 2 | 1266 | |||
XM_017016537.1 | XP_016872026.1 | 2 | 4862 | |||
XM_017016538.1 | XP_016872027.1 | 3 | 4278 | |||
XM_017016539.1 | XP_016872028.1 | 2 | 2005 | |||
XM_017016540.1 | XP_016872029.1 | 3 | 2131 | |||
XM_017016541.1 | XP_016872030.1 | 2 | 1567 | |||
XM_017016542.1 | XP_016872031.1 | 3 | 4386 | |||
XM_017016543.1 | XP_016872032.1 | 3 | 1091 | |||
GenBank mRNA | AK127465.1 | - | 4 | 1291 | ||
BC131825.1 | - | 1 | 776 | 40128007 | ||
BC137112.1 | - | 2 | 873 | 9021109 | ||
BC144478.1 | - | 2 | 873 | 9053005 |