Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAM196A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039762.2 | NP_001034851.1 | 4 | 1653 | ||
XM_005252694.3 | XP_005252751.1 | 2 | 1628 | |||
XM_017016537.1 | XP_016872026.1 | 2 | 5224 | |||
XM_017016538.1 | XP_016872027.1 | 3 | 4640 | |||
XM_017016539.1 | XP_016872028.1 | 2 | 2367 | |||
XM_017016540.1 | XP_016872029.1 | 3 | 2493 | |||
XM_017016541.1 | XP_016872030.1 | 2 | 1929 | |||
XM_017016542.1 | XP_016872031.1 | 3 | 4748 | |||
XM_017016543.1 | XP_016872032.1 | 3 | 1453 | |||
GenBank mRNA | AK127465.1 | - | 4 | 1653 | ||
BC131825.1 | - | 1 | 1138 | 40128007 | ||
BC137112.1 | - | 2 | 1235 | 9021109 | ||
BC144478.1 | - | 2 | 1235 | 9053005 |