Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080518.1 | NP_001073987.1 | 5 | 654 | ||
XM_011540073.2 | XP_011538375.1 | 6 | 755 | |||
XM_011540075.1 | XP_011538377.1 | 7 | 809 | |||
XM_011540077.1 | XP_011538379.1 | 6 | 746 | |||
XM_011540082.1 | XP_011538384.1 | 3 | 754 | |||
XM_017016545.1 | XP_016872034.1 | 6 | 755 | |||
XM_017016546.1 | XP_016872035.1 | 6 | 657 | |||
XM_017016547.1 | XP_016872036.1 | 6 | 756 | |||
XM_017016548.1 | XP_016872037.1 | 6 | 758 | |||
GenBank mRNA | EF426482.1 | - | 5 | 654 |