Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022475.2 | NP_071920.1 | 9 | 2154 | ||
XM_005263178.4 | XP_005263235.1 | 9 | 2023 | |||
XM_006714288.3 | XP_006714351.1 | 9 | 2023 | |||
GenBank mRNA | AK024645.1 | - | 2 | 141 | ||
AK291849.1 | - | 9 | 1494 | |||
AY009317.1 | - | 9 | 1994 | |||
AY009951.1 | - | 9 | 2129 | |||
AY358747.1 | - | 9 | 2104 | |||
BC025311.1 | - | 9 | 1980 | 4122618 |