Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039999.2 | NP_001035088.2 | 6 | 2586 | ||
XM_017024953.1 | XP_016880442.1 | 6 | 2670 | |||
GenBank mRNA | BC105988.1 | - | 2 | 570 | ||
CR735308.1 | - | 1 | 145 | |||
CX165458.1 | - | 2 | 373 |