Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085474.1 | NP_001078943.1 | 7 | 869 | ||
NM_001291283.1 | NP_001278212.1 | 6 | 721 | |||
GenBank mRNA | AY358469.1 | - | 7 | 821 | ||
BQ027877.1 | - | 1 | 333 |