Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM227A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013647.1 | NP_001013669.1 | 15 | 2082 | ||
NM_001291030.1 | NP_001277959.1 | 15 | 2122 | |||
XM_006724291.2 | XP_006724354.2 | 15 | 2094 | |||
XM_006724292.2 | XP_006724355.2 | 15 | 2082 | |||
XM_006724293.2 | XP_006724356.1 | 15 | 2094 | |||
XM_006724294.2 | XP_006724357.2 | 15 | 2134 | |||
XM_006724295.2 | XP_006724358.2 | 15 | 2122 | |||
XM_011530308.2 | XP_011528610.1 | 15 | 1799 | |||
XM_011530309.2 | XP_011528611.1 | 15 | 2094 | |||
XM_011530310.1 | XP_011528612.1 | 14 | 1989 | |||
XM_011530311.1 | XP_011528613.1 | 14 | 1971 | |||
XM_011530312.1 | XP_011528614.1 | 13 | 1371 | |||
XM_011530313.2 | XP_011528615.1 | 13 | 1323 | |||
XM_011530314.1 | XP_011528616.1 | 14 | 2024 | |||
GenBank mRNA | AK301819.1 | - | 15 | 1796 |