Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042574.2 | NP_001036039.1 | 10 | 1125 | ||
NM_022769.4 | NP_073606.3 | 10 | 1125 | |||
XM_005254968.2 | XP_005255025.1 | 10 | 1093 | |||
XM_011521906.2 | XP_011520208.1 | 9 | 997 | |||
GenBank mRNA | AK025521.1 | - | 3 | 292 | ||
AK074170.1 | - | 7 | 596 | |||
AK090443.1 | - | 10 | 1085 | |||
AK225300.1 | - | 3 | 292 | |||
AY360173.1 | - | 10 | 946 | |||
BC074730.2 | - | 1 | 68 | |||
BC074731.2 | - | 1 | 68 | 30915456 | ||
BP238198.1 | - | 3 | 293 | |||
DB288138.1 | - | 1 | 131 |