Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022772.3 | NP_073609.2 | 15 | 1605 | ||
XM_017018130.1 | XP_016873619.1 | 16 | 1822 | |||
XM_017018131.1 | XP_016873620.1 | 15 | 1693 | |||
XM_017018132.1 | XP_016873621.1 | 16 | 1663 | |||
GenBank mRNA | AF318331.1 | - | 3 | 774 | ||
AK025588.1 | - | 15 | 1523 | |||
AK025824.1 | - | 15 | 1523 | |||
AK027765.1 | - | 3 | 417 | |||
AK094539.1 | - | 7 | 701 | |||
AK122903.1 | - | 12 | 1801 | |||
AK222903.1 | - | 15 | 1518 | |||
AK225311.1 | - | 15 | 1523 | |||
AY074929.1 | - | 15 | 1630 | |||
BC080636.1 | - | 15 | 1609 | 6572993 | ||
BC093878.1 | - | 14 | 1398 | 7939723 | ||
BC101481.1 | - | 14 | 1398 | 8068987 | ||
BC143242.1 | - | 14 | 1446 | 9051743 |