Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511175.2 | XP_011509477.1 | 2 | 131 | ||
XM_011511177.2 | XP_011509479.1 | 2 | 131 |