Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256534.1 | NP_001243463.1 | 8 | 1093 | ||
NM_001287387.1 | NP_001274316.1 | 9 | 947 | |||
NM_005984.4 | NP_005975.1 | 9 | 1112 | |||
NR_046298.2 | 8 | 973 | ||||
GenBank mRNA | AK292313.1 | - | 9 | 1016 | ||
AK298204.1 | - | 9 | 945 | |||
BC004980.1 | - | 9 | 973 | 2957917 | ||
BC008061.2 | - | 9 | 975 | 2958945 | ||
BC018590.1 | - | 9 | 870 | |||
BM679710.1 | - | 1 | 619 | |||
HM037273.1 | - | 8 | 948 | |||
L75823.1 | - | 9 | 1021 | |||
L77567.1 | - | 9 | 994 | |||
U25147.1 | - | 9 | 996 |