Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003105.5 | NP_003096.1 | 29 | 4143 | ||
XM_011542963.2 | XP_011541265.1 | 28 | 3904 | |||
XM_011542965.2 | XP_011541267.1 | 23 | 3052 | |||
XM_011542967.2 | XP_011541269.1 | 9 | 1084 | |||
XM_017018169.1 | XP_016873658.1 | 29 | 3850 | |||
XM_017018170.1 | XP_016873659.1 | 27 | 3526 | |||
XM_017018171.1 | XP_016873660.1 | 29 | 4018 | |||
XM_017018172.1 | XP_016873661.1 | 15 | 2015 | |||
GenBank mRNA | AK295090.1 | - | 6 | 933 | ||
AK295878.1 | - | 9 | 1084 | |||
BC137171.1 | - | 29 | 4066 | 9021168 | ||
Y08110.1 | - | 29 | 4094 |