Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST5 SIRNA Assays ›
Gene Symbol:
ST5
Gene Name:
suppression of tumorigenicity 5
Gene Aliases:
DENND2B, HTS1, p126
Chromosome Location:
Chr.11: 8693352 - 8910951 on Build GRCh38
UniGene ID:
Hs.117715
Species:
Human
siRNA ID
HSS186122
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005418.3
NP_005409.3
7
1812
NM_139157.2
NP_631896.1
3
280
NM_213618.1
NP_998783.1
4
1612
XM_005253077.1
XP_005253134.1
4
1527
XM_005253083.2
XP_005253140.1
3
422
XM_011520309.1
XP_011518611.1
4
1527
XM_011520310.1
XP_011518612.1
4
1939
XM_011520311.2
XP_011518613.1
4
1921
XM_011520312.1
XP_011518614.1
8
1886
XM_011520313.1
XP_011518615.1
7
1772
XM_011520314.1
XP_011518616.1
6
1664
XM_011520315.1
XP_011518617.1
6
1760
XM_011520316.1
XP_011518618.1
5
1690
XM_011520317.1
XP_011518619.1
4
1631
XM_011520318.1
XP_011518620.1
8
1886
XM_011520319.2
XP_011518621.1
4
1479
XM_011520320.1
XP_011518622.1
4
1527
XM_011520321.2
XP_011518623.1
4
1527
XM_011520322.1
XP_011518624.1
4
1527
XM_011520323.2
XP_011518625.2
3
350
XM_011520324.2
XP_011518626.2
3
350
XM_011520325.1
XP_011518627.1
3
422
XM_011520326.1
XP_011518628.1
7
626
XM_011520327.1
XP_011518629.1
3
315
XM_011520328.1
XP_011518630.1
3
347
XM_011520329.1
XP_011518631.1
7
626
XM_017018182.1
XP_016873671.1
5
1645
XM_017018183.1
XP_016873672.1
5
1622
XM_017018184.1
XP_016873673.1
7
1781
XM_017018185.1
XP_016873674.1
3
1577
XM_017018186.1
XP_016873675.1
3
1577
XM_017018187.1
XP_016873676.1
3
286
GenBank mRNA
AK302824.1
-
3
259
AK312305.1
-
4
1578
AK312758.1
-
3
622
BC036655.1
-
7
1817
4837231
BX647431.1
-
4
1612
U15131.1
-
4
1540
U15780.1
-
3
280
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of GTPase activity
Rab guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0