Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193533.1 | NP_001180462.1 | 10 | 1787 | ||
NM_003157.4 | NP_003148.2 | 11 | 2054 | |||
XM_006713310.2 | XP_006713373.1 | 10 | 1913 | |||
XM_011534039.2 | XP_011532341.1 | 11 | 2051 | |||
XM_011534040.2 | XP_011532342.1 | 11 | 2051 | |||
XM_017007085.1 | XP_016862574.1 | 11 | 2051 | |||
XM_017007086.1 | XP_016862575.1 | 10 | 1913 | |||
GenBank mRNA | AK294165.1 | - | 10 | 1694 | ||
AK312420.1 | - | 11 | 2042 | |||
BC063044.1 | - | 10 | 1906 | 5169184 | ||
EF560744.1 | - | 11 | 2050 | |||
KJ592714.1 | - | 10 | 1713 | |||
L20321.1 | - | 11 | 2029 |