Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003192.2 | NP_003183.1 | 1 | 680 | ||
GenBank mRNA | AK312681.1 | - | 1 | 622 | ||
BC017479.1 | - | 2 | 1171 | 3357283 | ||
BC020170.1 | - | 1 | 620 | 3536688 | ||
BE795945.1 | - | 1 | 680 | |||
BT007002.1 | - | 1 | 607 | |||
U61234.1 | - | 1 | 630 |