Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
siRNA ID
HSS186230
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
10
1377
NM_001243226.2
NP_001230155.2
11
1167
NM_001243227.1
NP_001230156.1
9
1211
NM_001243228.1
NP_001230157.1
10
1321
NM_001243230.1
NP_001230159.1
9
853
NM_001243231.1
NP_001230160.1
8
811
NM_001243232.1
NP_001230161.1
6
833
NM_001243233.1
NP_001230162.1
6
774
NM_001243234.1
NP_001230163.1
3
406
NM_001243235.1
NP_001230164.1
3
406
NM_001243236.1
NP_001230165.1
3
422
NM_001306207.1
NP_001293136.1
9
1002
NM_001306208.1
NP_001293137.1
6
833
NM_003199.2
NP_003190.1
10
1377
XM_005266739.3
XP_005266796.2
9
1017
XM_005266741.3
XP_005266798.1
10
977
XM_005266743.4
XP_005266800.1
9
838
XM_005266744.3
XP_005266801.1
9
958
XM_005266745.3
XP_005266802.1
9
820
XM_005266747.3
XP_005266804.1
8
814
XM_005266749.3
XP_005266806.1
6
605
XM_005266752.4
XP_005266809.1
6
1175
XM_005266754.3
XP_005266811.1
5
599
XM_005266755.4
XP_005266812.1
6
634
XM_005266761.3
XP_005266818.1
3
918
XM_006722536.2
XP_006722599.1
10
1300
XM_006722537.2
XP_006722600.1
10
933
XM_006722538.2
XP_006722601.1
9
955
XM_011526158.2
XP_011524460.1
9
1931
XM_011526160.2
XP_011524462.1
5
984
XM_017025934.1
XP_016881423.1
9
1014
XM_017025935.1
XP_016881424.1
9
1017
XM_017025936.1
XP_016881425.1
9
1017
XM_017025937.1
XP_016881426.1
9
771
XM_017025938.1
XP_016881427.1
10
930
XM_017025939.1
XP_016881428.1
10
980
XM_017025940.1
XP_016881429.1
10
1300
XM_017025941.1
XP_016881430.1
10
933
XM_017025942.1
XP_016881431.1
10
977
XM_017025943.1
XP_016881432.1
10
980
XM_017025944.1
XP_016881433.1
9
1235
XM_017025945.1
XP_016881434.1
9
1238
XM_017025946.1
XP_016881435.1
9
1252
XM_017025947.1
XP_016881436.1
9
838
XM_017025948.1
XP_016881437.1
9
1235
XM_017025949.1
XP_016881438.1
9
955
XM_017025950.1
XP_016881439.1
9
1235
XM_017025951.1
XP_016881440.1
8
814
XM_017025952.1
XP_016881441.1
6
828
XM_017025953.1
XP_016881442.1
6
605
XM_017025954.1
XP_016881443.1
6
1189
XM_017025955.1
XP_016881444.1
5
923
XM_017025956.1
XP_016881445.1
5
599
XM_017025957.1
XP_016881446.1
3
918
GenBank mRNA
AB209741.1
-
5
485
AK095041.1
-
6
833
AK096862.1
-
8
811
AK122765.1
-
10
1377
AK299169.1
-
9
1211
AK300038.1
-
3
352
AK300612.1
-
6
774
AK300636.1
-
5
504
AK301144.1
-
9
988
AK315074.1
-
10
1309
AK316165.1
-
5
548
AL518903.3
-
3
422
BC031056.1
-
11
1128
BC125084.1
-
9
791
40117795
BC125085.1
-
9
791
40117798
DC298029.1
-
3
406
FR748210.1
-
9
765
FR748211.1
-
9
765
FR748212.1
-
7
585
FR748213.1
-
7
585
FR748214.1
-
8
693
FR748215.1
-
6
513
FR748216.1
-
9
759
FR748217.1
-
8
639
FR748218.1
-
4
375
FR748219.1
-
6
552
FR748220.1
-
3
285
FR748221.1
-
3
285
FR748222.1
-
3
285
FR748223.1
-
1
117
M74718.1
-
3
237
M74719.1
-
10
964
X52079.1
-
7
630
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0