Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017489.2 | NP_059523.2 | 7, 8 | 960 | ||
XM_005251291.3 | XP_005251348.1 | 7, 8 | 958 | |||
XM_011517582.2 | XP_011515884.1 | 8, 9 | 1025 | |||
GenBank mRNA | AK292529.1 | - | 7, 8 | 958 | ||
U40705.1 | - | 7, 8 | 950 |