Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: THOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003249.4 | NP_003240.1 | 10, 11 | 1853 | ||
XM_011528228.2 | XP_011526530.1 | 7, 8 | 2800 | |||
GenBank mRNA | AK093392.1 | - | 1, 2 | 971 | ||
AK300552.1 | - | 7, 8 | 1335 | |||
BC000135.2 | - | 10, 11 | 1779 | 2900551 | ||
BC000583.2 | - | 8, 9 | 1358 | |||
BC002391.2 | - | 10, 11 | 1805 | 2819803 | ||
BC013878.2 | - | 10, 11 | 1769 | 3690133 | ||
U29366.1 | - | 10, 11 | 1737 | |||
Z50115.1 | - | 10, 11 | 1777 |