Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003249.4 | NP_003240.1 | 6 | 835 | ||
XM_011528228.2 | XP_011526530.1 | 3 | 1782 | |||
GenBank mRNA | AK300552.1 | - | 3 | 317 | ||
BC000135.2 | - | 6 | 761 | 2900551 | ||
BC000583.2 | - | 4 | 340 | |||
BC002391.2 | - | 6 | 787 | 2819803 | ||
BC013878.2 | - | 6 | 751 | 3690133 | ||
U29366.1 | - | 6 | 719 | |||
Z50115.1 | - | 6 | 759 |