Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006289.3 | NP_006280.3 | 11 | 1532 | ||
GenBank mRNA | AB028950.2 | - | 11 | 1507 | ||
AF078828.1 | - | 11 | 1304 | |||
AF177198.1 | - | 11 | 1268 | |||
BC042923.1 | - | 11 | 1318 | 5576203 |