Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM7SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277233.1 | NP_001264162.1 | 6 | 828 | ||
NM_003273.3 | NP_003264.2 | 6 | 828 | |||
NR_102367.1 | 5 | 773 | ||||
GenBank mRNA | AF023676.1 | - | 4 | 1322 | ||
AF096304.1 | - | 6 | 792 | |||
AK290935.1 | - | 6 | 806 | |||
AK296441.1 | - | 5 | 749 | |||
BC009052.1 | - | 6 | 794 | 3874367 | ||
BC012857.1 | - | 6 | 756 | 4214662 | ||
BC038353.1 | - | 6 | 798 | 3861907 |