Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM3 SIRNA Assays ›
Gene Symbol:
TPM3
Gene Name:
tropomyosin 3
Gene Aliases:
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30
Chromosome Location:
Chr.1: 154155304 - 154192135 on Build GRCh38
UniGene ID:
Hs.535581
Species:
Human
siRNA ID
HSS186399
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001043351.1
NP_001036816.1
6
698
NM_001043352.1
NP_001036817.1
6
698
NM_001043353.1
NP_001036818.1
6
698
NM_001278188.1
NP_001265117.1
5
564
NM_001278189.1
NP_001265118.1
6
698
NM_001278191.1
NP_001265120.1
6
544
NM_152263.3
NP_689476.2
7
796
NM_153649.3
NP_705935.1
6
698
NR_103461.1
6
698
GenBank mRNA
AB062125.1
-
2
111
AF474157.1
-
6
568
AK026559.1
-
6
638
AK056889.1
-
2
687
AK056921.1
-
6
555
AK056997.1
-
7
586
AK092690.1
-
7
636
AK092712.1
-
5
443
AK123571.1
-
1
1344
AK125367.1
-
3
2582
AK298678.1
-
5
542
AK301674.1
-
6
520
AK308007.1
-
5
473
AK315504.1
-
6
657
AL537561.3
-
6
608
AY004867.1
-
6
579
BC000771.2
-
6
610
3506357
BC008407.1
-
7
762
4245271
BC008425.1
-
7
765
4246114
BC015403.1
-
6
613
4389911
BC062740.1
-
7
628
4271142
BC072428.1
-
6
625
6472532
BF967161.1
-
6
512
BG706843.1
-
6
641
BI546300.1
-
6
609
BM674269.1
-
4
626
BU956481.1
-
6
624
BX419658.2
-
6
610
BX648485.1
-
6
610
CR997359.1
-
6
589
EU668324.1
-
7
636
FJ224335.1
-
6
610
KU178368.1
-
6
568
KU178369.1
-
6
568
KU178370.1
-
6
568
M12128.1
-
1
196
X04201.1
-
7
737
X04588.1
-
6
620
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
muscle contraction
muscle filament sliding
molecular_function
actin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0