Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000064.3 | NP_000055.2 | 6 | 759 | ||
GenBank mRNA | AK304071.1 | - | 6 | 727 | ||
BC150179.1 | - | 6 | 698 | 40148091 | ||
BC150200.1 | - | 6 | 698 | 40148812 | ||
EU794602.1 | - | 6 | 698 | |||
K02765.1 | - | 6 | 726 |