Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007118.3 | NP_009049.2 | 42 | 6242 | ||
NR_134469.1 | 42 | 6242 | ||||
XM_011514107.2 | XP_011512409.1 | 42 | 6817 | |||
XM_011514108.1 | XP_011512410.1 | 42 | 6116 | |||
XM_011514109.2 | XP_011512411.1 | 42 | 9894 | |||
XM_011514110.2 | XP_011512412.1 | 42 | 6352 | |||
XM_017009801.1 | XP_016865290.1 | 42 | 6594 | |||
XM_017009802.1 | XP_016865291.1 | 42 | 6594 | |||
XM_017009803.1 | XP_016865292.1 | 35 | 4863 | |||
GenBank mRNA | AB115332.1 | - | 6 | 506 | ||
AB209754.1 | - | 37 | 5136 | |||
AF091395.1 | - | 42 | 6107 | |||
U42390.1 | - | 42 | 6107 |