Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MROH1 SIRNA Assays ›
Gene Symbol:
MROH1
Gene Name:
maestro heat like repeat family member 1
Gene Aliases:
HEATR7A
Chromosome Location:
Chr.8: 144148016 - 144261940 on Build GRCh38
UniGene ID:
Hs.443139
Species:
Human
siRNA ID
HSS186541
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MROH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099280.2
NP_001092750.1
11
1241
NM_001099281.2
NP_001092751.1
10
1121
NM_001288814.1
NP_001275743.1
10
1128
NM_032450.2
NP_115826.2
11
1241
XM_011517270.2
XP_011515572.1
11
1379
XM_011517271.2
XP_011515573.1
11
1378
XM_011517272.2
XP_011515574.1
11
1379
XM_011517273.2
XP_011515575.1
11
1377
XM_011517274.2
XP_011515576.1
11
1377
XM_011517275.2
XP_011515577.1
11
1377
XM_011517276.2
XP_011515578.1
11
1405
XM_011517277.1
XP_011515579.1
11
1241
XM_011517278.1
XP_011515580.1
10
1121
XM_011517279.2
XP_011515581.1
10
1285
XM_011517281.1
XP_011515583.1
10
1294
XM_011517282.1
XP_011515584.1
9
1219
XM_011517283.2
XP_011515585.1
11
1200
XM_011517285.1
XP_011515587.1
10
1121
XM_011517286.2
XP_011515588.1
11
1373
XM_017013802.1
XP_016869291.1
11
1377
XM_017013803.1
XP_016869292.1
11
1376
XM_017013804.1
XP_016869293.1
11
1241
XM_017013805.1
XP_016869294.1
10
1121
XM_017013806.1
XP_016869295.1
8
856
XM_017013807.1
XP_016869296.1
11
1374
GenBank mRNA
AB058736.1
-
10
1133
AK295740.1
-
10
1121
AL834309.1
-
10
1125
BC040742.1
-
8
868
BC057824.1
-
11
1221
BC067855.1
-
11
1221
BC092486.1
-
10
1101
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0