Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142308.2 | NP_001135780.2 | 18 | 2965 | ||
XM_011519453.2 | XP_011517755.1 | 19 | 3503 | |||
XM_011519454.1 | XP_011517756.1 | 19 | 2829 | |||
XM_011519455.2 | XP_011517757.1 | 18 | 3330 | |||
XM_017016182.1 | XP_016871671.1 | 18 | 2775 | |||
XM_017016183.1 | XP_016871672.1 | 19 | 3504 | |||
XM_017016184.1 | XP_016871673.1 | 7 | 1192 | |||
XM_017016185.1 | XP_016871674.1 | 7 | 1189 | |||
GenBank mRNA | KC843478.1 | - | 18 | 2965 |