Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101417.3 | NP_001094887.1 | 7 | 925 | ||
NM_001101426.3 | NP_001094896.1 | 8 | 1075 | |||
XM_006715770.3 | XP_006715833.1 | 8 | 1245 | |||
XM_011515499.2 | XP_011513801.1 | 8 | 1220 | |||
XM_011515500.2 | XP_011513802.1 | 7 | 1115 | |||
XM_011515508.2 | XP_011513810.1 | 8 | 1577 | |||
XM_011515509.2 | XP_011513811.1 | 8 | 2426 | |||
XM_017012575.1 | XP_016868064.1 | 8 | 1220 | |||
XM_017012577.1 | XP_016868066.1 | 7 | 1337 | |||
XM_017012578.1 | XP_016868067.1 | 6 | 1752 | |||
GenBank mRNA | BE092109.1 | - | 1 | 16 | ||
DY654823.1 | - | 5 | 328 |