Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256648.1 | NP_001243577.1 | 7 | 1326 | ||
NM_001256649.1 | NP_001243578.1 | 7 | 1191 | |||
NM_001256650.1 | NP_001243579.1 | 6 | 1063 | |||
NM_001256651.1 | NP_001243580.1 | 3 | 677 | |||
NM_001256653.1 | NP_001243582.1 | 4 | 804 | |||
NM_001256654.1 | NP_001243583.1 | 3 | 686 | |||
NM_003423.3 | NP_003414.2 | 4 | 813 | |||
XM_011528257.2 | XP_011526559.1 | 4 | 735 | |||
XM_011528259.2 | XP_011526561.1 | 4 | 1292 | |||
XM_017027207.1 | XP_016882696.1 | 7 | 6804 | |||
XM_017027208.1 | XP_016882697.1 | 7 | 1484 | |||
XM_017027209.1 | XP_016882698.1 | 7 | 6537 | |||
XM_017027210.1 | XP_016882699.1 | 6 | 6328 | |||
XM_017027211.1 | XP_016882700.1 | 7 | 6471 | |||
XM_017027212.1 | XP_016882701.1 | 6 | 1331 | |||
XM_017027213.1 | XP_016882702.1 | 7 | 6579 | |||
XM_017027214.1 | XP_016882703.1 | 5 | 1254 | |||
XM_017027215.1 | XP_016882704.1 | 6 | 1093 | |||
XM_017027216.1 | XP_016882705.1 | 5 | 879 | |||
GenBank mRNA | AF112859.1 | - | 1 | 230 | ||
AK291353.1 | - | 7 | 1141 | |||
BC006528.2 | - | 4 | 780 | 3687640 | ||
BT009756.1 | - | 3 | 625 | |||
CR933640.1 | - | 7 | 1286 | |||
X59244.1 | - | 6 | 962 |