Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085399.1 | NP_001078868.1 | 5, 6 | 761 | ||
NM_001085400.1 | NP_001078869.1 | 5, 6 | 761 | |||
XM_017008590.1 | XP_016864079.1 | 3, 4 | 429 | |||
GenBank mRNA | AK075468.1 | - | 5, 6 | 760 | ||
BC039540.1 | - | 5, 6 | 759 | 5748847 | ||
DA491231.1 | - | 1, 2 | 127 |